EMAIL THIS PAGE TO A FRIEND
8.22117 EMD Millipore

Ethyl oxamate

Ethyl oxamate for synthesis. CAS 617-36-7, EC Number 210-512-8, chemical formula C₂H₅OOCCONH₂., for synthesis

Synonym: Oxalic acid monoethyl ester amide, Oxamic acid ethyl ester, Oxamethane

  • CAS Number 617-36-7

  • Linear Formula C₄H₇NO₃

  • Molecular Weight 117.1 g/mol

  •  EC Number 210-512-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8221170025)
mp   112 - 114 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (ex N): ≥ 98.0 %
  Melting range (lower value): ≥ 114 °C
  Melting range (upper value): ≤ 116 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 19 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?