• USA Home
  • 8.22118 - Ethyl 2-oxocyclohexanecarboxylate

EMAIL THIS PAGE TO A FRIEND
8.22118 EMD Millipore

Ethyl 2-oxocyclohexanecarboxylate

Ethyl 2-oxocyclohexanecarboxylate for synthesis. CAS 1655-07-8, chemical formula 2-(O=)C₆H₉COOC₂H₅., for synthesis

Synonym: 2-Oxocyclohexanecarboxylic acid ethyl ester, Cyclohexanone-2-carboxylic acid ethyl ester, 2-Carbethoxycyclohexanone

  • CAS Number 1655-07-8

  • Linear Formula C₉H₁₄O₃

  • Molecular Weight 170.2 g/mol

  •  EC Number 216-731-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8221180100)
bp   106 - 107 °C (15 hPa)
density   1.071 g/cm3 (20 °C)
flash point   101 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.069 - 1.074
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 30 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?