• USA Home
  • 8.22124 - Phenylacetaldehyde dimethyl acetal

EMAIL THIS PAGE TO A FRIEND
8.22124 EMD Millipore

Phenylacetaldehyde dimethyl acetal

Phenylacetaldehyde dimethyl acetal for synthesis. CAS 101-48-4, EC Number 202-945-6, chemical formula C₆H₅CH₂CH(OCH₃)₂., for synthesis

Synonym: 1,1-Dimethoxy-2-phenylethane

  • CAS Number 101-48-4

  • Linear Formula C₁₀H₁₄O₂

  • Molecular Weight 166.21 g/mol

  •  EC Number 202-945-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8221240250)
bp   110 °C (29 hPa)
density   1.00 g/cm3 (20 °C)
flash point   89 °C
LD 50 oral   LD50 Rat 3500 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.003 - 1.004
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2911 00 00
RTECS 
AB3040000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?