• USA Home
  • 8.22128 - Phenyl 2-pyridyl ketone oxime

EMAIL THIS PAGE TO A FRIEND
8.22128 EMD Millipore

Phenyl 2-pyridyl ketone oxime

Phenyl 2-pyridyl ketone oxime for synthesis. CAS 1826-28-4, EC Number 217-374-8, chemical formula 2-(C₆H₅C=NOH)C₅H₄N., for synthesis

  • CAS Number 1826-28-4

  • Linear Formula C₁₂H₁₀N₂O

  • Molecular Weight 198.23 g/mol

  •  EC Number 217-374-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8221280001)
mp   152 - 155 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 152 °C
  Melting range (upper value): ≤ 155 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?