• USA Home
  • 8.22168 - Methyl trichloroacetate

EMAIL THIS PAGE TO A FRIEND
8.22168 EMD Millipore

Methyl trichloroacetate

Methyl trichloroacetate for synthesis. CAS 598-99-2, EC Number 209-960-7, chemical formula CCl₃COOCH₃., for synthesis

Synonym: Trichloroacetic acid methyl ester

  • CAS Number 598-99-2

  • Linear Formula C₃H₃Cl₃O₂

  • Molecular Weight 177.41 g/mol

  •  EC Number 209-960-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8221680050)
solubility   1050 mg/l
density   1.49 g/cm3 (20 °C)
flash point   85 °C
mp   -17.5 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.485 - 1.487
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 40 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 2533 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2533 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2533 , 6.1, III
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
S Phrase 
S 26
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?