• USA Home
  • 8.22172 - 4-Aminobenzotrifluoride

EMAIL THIS PAGE TO A FRIEND
8.22172 EMD Millipore

4-Aminobenzotrifluoride

4-Aminobenzotrifluoride for synthesis. CAS 455-14-1, chemical formula 4-(NH₂)C₆H₄CF₃., for synthesis

Synonym: 4-(Trifluoromethyl)aniline, α,α,α-Trifluoro-p-toluidine

  • CAS Number 455-14-1

  • Linear Formula C₇H₆F₃N

  • Molecular Weight 161.12 g/mol

  •  EC Number 207-236-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8221720005)
bp   82 - 84 °C (16 hPa)
vapor pressure   1 hPa (38 °C)
density   1.30 g/cm3 (20 °C)
flash point   86 °C
mp   3 - 8 °C
LD 50 oral   LD50 Rat 128 mg/kg
LD 50 dermal   LD50 Rat > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.294 - 1.297
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 43 00
Hazard Statement(s) 
H301: Toxic if swallowed.
H318: Causes serious eye damage.
H410: Very toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear eye protection.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
XU9260000
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2810 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2810 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2810 , 6.1, III
Categories of danger 
toxic, irritant, dangerous for the environment
R Phrase 
R 25-41-50/53
Toxic if swallowed.Risk of serious damage to eyes.Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 26-37/39-45-61
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?