• USA Home
  • 8.22175 - 2,4,6-Trimethylbenzaldehyde

EMAIL THIS PAGE TO A FRIEND
8.22175 EMD Millipore

2,4,6-Trimethylbenzaldehyde

2,4,6-Trimethylbenzaldehyde for synthesis. CAS 487-68-3, chemical formula 2,4,6-(CH₃)₃C₆H₂CHO., for synthesis

Synonym: Mesitaldehyde, Mesitylene-2-carbaldehyde

  • CAS Number 487-68-3

  • Linear Formula C₁₀H₁₂O

  • Molecular Weight 148.21 g/mol

  •  EC Number 207-662-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8221750010)
density   1.02 g/cm3 (20 °C)
flash point   105 °C
mp   14 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.018 - 1.019
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2912 29 00
Hazard Statement(s) 
H315: Causes skin irritation.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
RTECS 
CU8500000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 3082 , 9, III
Declaration (transport by air) IATA-DGR 
UN 3082 , 9, III
Declaration (transport by sea) IMDG-Code 
UN 3082 , 9, III, Marine Pollutant: P
R Phrase 
R 38-51/53
Irritating to skin.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 37-61
Wear suitable gloves.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?