• USA Home
  • 8.24499 - (R)-(-)-1,1'-Binaphthalene-2,2'-diyl-phosphate

EMAIL THIS PAGE TO A FRIEND
8.24499 EMD Millipore

(R)-(-)-1,1'-Binaphthalene-2,2'-diyl-phosphate

(R)-(-)-1,1'-Binaphthalene-2,2'-diyl-phosphate for synthesis. CAS 39648-67-4, molar mass 348.29 g/mol., for synthesis

Synonym: (R)-4-Hydroxy dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-oxide, BNPA, BPA

  • CAS Number 39648-67-4

  • Linear Formula C₂₀H₁₃O₄P

  • Molecular Weight 348.29 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 mg in Glass bottle (8244990250)
mp   >300 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 99.0 % (a/a)
  Spec. rotation [α²0/D (c=1 in methanol): -610.0 - -600.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2919 90 00
Hazard Statement(s) 
H315: Causes skin irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 38
Irritating to skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?