• USA Home
  • 8.24505 - (4R)-(+)-4-Isopropyl-2-oxazolidinone

EMAIL THIS PAGE TO A FRIEND
8.24505 EMD Millipore

(4R)-(+)-4-Isopropyl-2-oxazolidinone

(4R)-(+)-4-Isopropyl-2-oxazolidinone for synthesis. CAS 95530-58-8, molar mass 129.16 g/mol., for synthesis

  • CAS Number 95530-58-8

  • Linear Formula C₆H₁₁NO₂

  • Molecular Weight 129.16 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks
packaging   250 mg in Glass bottle (8245050250)
vapor pressure   0.0026 hPa (25 °C)
density   1.04 g/cm3
flash point   129.9 °C
mp   70 - 73 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.5 % (a/a)
  Melting range (lower value): ≥ 70 °C
  Melting range (upper value): ≤ 73 °C
  Spec. rotation [α²0/D (c=1 in methanol): +22.0 - +24.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 99 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?