• USA Home
  • 8.40044 - Kryptofix® 22 polymer (Merrifield)

EMAIL THIS PAGE TO A FRIEND
8.40044 EMD Millipore

Kryptofix® 22 polymer (Merrifield)

for synthesis

Synonym: 1,4,10,13-Tetraoxa-7,16-diazacyclooctadecane

Purchase

Properties

packaging   1 g in Glass bottle (8400440001)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay of N: 3.0 - 5.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 99 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?