• USA Home
  • 8.40049 - O-(2-Chlorobenzyloxycarbonyl)-N-hydroxysuccinimide

EMAIL THIS PAGE TO A FRIEND
8.40049 EMD Millipore

O-(2-Chlorobenzyloxycarbonyl)-N-hydroxysuccinimide

O-(2-Chlorobenzyloxycarbonyl)-N-hydroxysuccinimide for synthesis. CAS 65853-65-8, molar mass 283.67 g/mol., for synthesis

  • CAS Number 65853-65-8

  • Linear Formula C₁₂H₁₀ClNO₅

  • Molecular Weight 283.67 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 g in Glass bottle (8400490050)
mp   101 - 105 °C
LD 50 oral   LD50 Rat > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 94.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2929 90 00
Hazard Statement(s) 
H317: May cause an allergic skin reaction.
Precautionary Statement(s) 
P280: Wear protective gloves.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
sensitizing
R Phrase 
R 43
May cause sensitisation by skin contact.
S Phrase 
S 24-37
Avoid contact with skin.Wear suitable gloves.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?