• USA Home
  • 8.40066 - 2-Methylglutarodinitrile

EMAIL THIS PAGE TO A FRIEND
8.40066 EMD Millipore

2-Methylglutarodinitrile

2-Methylglutarodinitrile for synthesis. CAS 4553-62-2, molar mass 108.14 g/mol., for synthesis

Synonym: 1,3-Dicyanobutane, 2-Methylpentanedinitrile

  • CAS Number 4553-62-2

  • Linear Formula C₆H₈N₂

  • Molecular Weight 108.14 g/mol

  •  EC Number 224-923-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8400660050)
expl. lim.     0.3 - 3.25 %(V)
vapor pressure   0.007 hPa (25 °C)
solubility   40 g/l
density   0.953 g/cm3
flash point   115 °C
autoignition temp.   538 °C
mp   -45 °C
LD 50 oral   LD50 Rat 205 mg/kg
LD 50 dermal   LD50 Rat 1154 mg/kg
shipped in   ambient
storage temp.   Store at +2°C to +8°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.950 - 0.954
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Hazard Statement(s) 
H301: Toxic if swallowed.
H312: Harmful in contact with skin.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
YV8140000
Storage class 
6.1 C Combustible, acute toxicity cat. 3 / toxic or substances with chronic effects
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 3276 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 3276 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 3276 , 6.1, III
Categories of danger 
harmful
R Phrase 
R 21/22
Harmful in contact with skin and if swallowed.
S Phrase 
S 25-36/37
Avoid contact with eyes.Wear suitable protective clothing and gloves.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?