• USA Home
  • 8.40089 - (Methacryloyloxyethyl)trimethyl ammonium chloride

EMAIL THIS PAGE TO A FRIEND
8.40089 EMD Millipore

(Methacryloyloxyethyl)trimethyl ammonium chloride

(75% solution in water) (stabilised with hydroquinone monomethyl ether) for synthesis

Purchase

Properties

density   1.10 g/cm3 (20 °C)
LD 50 oral   LD50 Rat > 2000 mg/kg
LD 50 dermal   LD50 Rat > 2000 mg/kg
shipped in   ambient
storage temp.   Store at +2°C to +8°C.
specifications   Assay (argentometric): ≥ 74.0 %
  Density (d 20 °C/ 4 °C): 1.100 - 1.110
  Water (K. F.): ≤ 26.0 %

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2923 90 00
Hazard Statement(s) 
H317: May cause an allergic skin reaction.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P280: Wear protective gloves.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Categories of danger 
irritant, sensitizing
R Phrase 
R 36-43
Irritating to eyes.May cause sensitisation by skin contact.
S Phrase 
S 24-37
Avoid contact with skin.Wear suitable gloves.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?