• USA Home
  • 8.40095 - 3-Chlorobenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
8.40095 EMD Millipore

3-Chlorobenzyl alcohol

3-Chlorobenzyl alcohol for synthesis. CAS 873-63-2, molar mass 142.58 g/mol., for synthesis

  • CAS Number 873-63-2

  • Linear Formula C₇H₇ClO

  • Molecular Weight 142.58 g/mol

  •  EC Number 212-847-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8400950005)
bp   237 °C
density   1.22 g/cm3
flash point   110 °C
refractive index   1.5521 (25 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.220 - 1.225
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 29 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?