• USA Home
  • 8.40096 - 2-Methyl-1-phenyl-2-propanol

EMAIL THIS PAGE TO A FRIEND
8.40096 EMD Millipore

2-Methyl-1-phenyl-2-propanol

2-Methyl-1-phenyl-2-propanol for synthesis. CAS 100-86-7, molar mass 150.22 g/mol., for synthesis

Synonym: α,α-Dimethyl phenethyl alcohol, Dimethyl benzyl carbinol

  • CAS Number 100-86-7

  • Linear Formula C₁₀H₁₄O

  • Molecular Weight 150.22 g/mol

  •  EC Number 202-896-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   500 ml in Glass bottle (8400960500)
bp   215 °C
density   0.974 g/cm3 (20 °C)
flash point   82 °C
mp   23 - 25 °C
LD 50 oral   LD50 Rat 1280 mg/kg
LD 50 dermal   LD50 Rabbit > 5000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.972 - 0.976
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 29 00
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?