• USA Home
  • 8.40103 - 2-Amino-4-chloro-diphenyl ether

EMAIL THIS PAGE TO A FRIEND
8.40103 EMD Millipore

2-Amino-4-chloro-diphenyl ether

2-Amino-4-chloro-diphenyl ether for synthesis. CAS 93-67-4, molar mass 219.67 g/mol., for synthesis

Synonym: 5-Chloro-2-phenoxyaniline

  • CAS Number 93-67-4

  • Linear Formula C₁₂H₁₀ClNO

  • Molecular Weight 219.67 g/mol

  •  EC Number 202-266-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8401030025)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 41 °C
  Melting range (upper value): ≤ 45 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?