EMAIL THIS PAGE TO A FRIEND
8.40128 EMD Millipore

Tin(II) oxalate

Tin(II) oxalate for synthesis. CAS 814-94-8, molar mass 206.73 g/mol., for synthesis

  • CAS Number 814-94-8

  • Linear Formula C₂O₄Sn

  • Molecular Weight 206.73 g/mol

  •  EC Number 212-414-0

Purchase

Properties

packaging   50 g in Plastic bottle (8401280050)
solubility   0.5 g/l
density   3.55 g/cm3 (20 °C)
autoignition temp.   550 °C
mp   280 °C (decomposition)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (iodometric): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 11 00
Hazard Statement(s) 
H302 + H312: Harmful if swallowed or in contact with skin
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
RTECS 
XQ3950000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
15
Heavy metal-containing solutions and solids: container E. Stir Raney nickel (also: Urushibara nickel) in the form of an aqueous suspension into hydrochloric acid (Cat. No. 100312) until dissolved (container E). Neither Raney nickel itself nor any filter residues should be allowed to dry out, otherwise they will spontaneously ignite in air. In this context, heavy metal means any compound of antimony, arsenic, cadmium, chromium(VI), copper, lead, nickel and tin, as well as these subtances in metallic form, if they are classified as hazardous (acc. to AbfallverzeichnisV - Waste Catalogue Ordinance, Appendix 3). Other heavy metals should be collected separately.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 21/22
Harmful in contact with skin and if swallowed.
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?