• USA Home
  • 8.40129 - Butyltinchloride-dihydroxide

EMAIL THIS PAGE TO A FRIEND
8.40129 EMD Millipore

Butyltinchloride-dihydroxide

Butyltinchloride-dihydroxide for synthesis. CAS 13355-96-9, molar mass 245.27 g/mol., for synthesis

Synonym: Butyl chloro dihydroxy stannan

  • CAS Number 13355-96-9

  • Linear Formula C₄H₁₁ClO₂Sn

  • Molecular Weight 245.27 g/mol

  •  EC Number 236-406-1

Purchase

Properties

packaging   10 g in Glass bottle (8401290010)
bp   >300 °C (1013 hPa)
vapor pressure   1.10 hPa (20 °C)
mp   144 °C
LD 50 oral   LD50 Rat 2459 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 2 water endangering
Disposal 
15
Heavy metal-containing solutions and solids: container E. Stir Raney nickel (also: Urushibara nickel) in the form of an aqueous suspension into hydrochloric acid (Cat. No. 100312) until dissolved (container E). Neither Raney nickel itself nor any filter residues should be allowed to dry out, otherwise they will spontaneously ignite in air. In this context, heavy metal means any compound of antimony, arsenic, cadmium, chromium(VI), copper, lead, nickel and tin, as well as these subtances in metallic form, if they are classified as hazardous (acc. to AbfallverzeichnisV - Waste Catalogue Ordinance, Appendix 3). Other heavy metals should be collected separately.
Declaration (railroad and road) ADR, RID 
UN 3146 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 3146 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 3146 , 6.1, III, Marine Pollutant: P
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?