• USA Home
  • 8.40131 - (R)-(+)-1-Phenylethanol

EMAIL THIS PAGE TO A FRIEND
8.40131 EMD Millipore

(R)-(+)-1-Phenylethanol

(R)-(+)-1-Phenylethanol for synthesis. CAS 1517-69-7, molar mass 122.16 g/mol., for synthesis

Synonym: (R)-(+)-Methyl phenyl carbinol

  • CAS Number 1517-69-7

  • Linear Formula C₈H₁₀O

  • Molecular Weight 122.16 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 ml in Glass bottle (8401310001)
bp   98 °C (27 hPa)
vapor pressure   13 Pa (20 °C)
density   1.012 g/cm3
flash point   85 °C
mp   9 - 11 °C
pH   7 (H₂O)
LD 50 oral   LD50 Rat 400 mg/kg
LD 50 dermal   LD50 Rabbit 2500 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.011 - 1.013
  Spec. rotation [α²0/D (undiluted): +40.0 - +44.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 29 00
Hazard Statement(s) 
H302: Harmful if swallowed.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P280: Wear eye protection.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2937 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2937 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2937 , 6.1, III
Categories of danger 
harmful, irritant
R Phrase 
R 22-36/38
Harmful if swallowed.Irritating to eyes and skin.
S Phrase 
S 24-26
Avoid contact with skin.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?