• USA Home
  • 8.40134 - (2-Hydroxyethyl)-salicylate

EMAIL THIS PAGE TO A FRIEND
8.40134 EMD Millipore

(2-Hydroxyethyl)-salicylate

(2-Hydroxyethyl)-salicylate for synthesis. CAS 87-28-5, pH 4.0 (13.1 g/l, H₂O, 20 °C)., for synthesis

Synonym: Salicylic acid (2-hydroxyethyl) ester

  • CAS Number 87-28-5

  • Linear Formula C₉H₁₀O₄

  • Molecular Weight 182.17 g/mol

  •  EC Number 201-737-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8401340100)
bp   150 - 155 °C (5 hPa)
vapor pressure   0.0001 hPa (20 °C)
solubility   13.1 g/l
density   1.25 g/cm3
flash point   171 °C DIN 51758
autoignition temp.   441 °C DIN 51794
mp   30 °C
pH   4.0 (13.1 g/l, H₂O, 20 °C)
LD 50 oral   LD50 Rat 1380 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 23 00
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?