• USA Home
  • 8.40143-M - 1,4-Butanediolmonovinylether

EMAIL THIS PAGE TO A FRIEND
8.40143-M EMD Millipore

1,4-Butanediolmonovinylether

1,4-Butanediolmonovinylether (stabilized with potassium hydroxide) for synthesis. CAS 17832-28-9, molar mass 116.15 g/mol., (stabilized with potassium hydroxide) for synthesis

  • CAS Number 17832-28-9

  • Linear Formula C₆H₁₂O₂

  • Molecular Weight 116.15 g/mol

  •  EC Number 241-793-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8401430050)
bp   189 °C (1013 hPa)
expl. lim.     1.4 - 9.9 %(V)
vapor pressure   0.3 hPa (20 °C)
solubility   75 g/l
density   0.94 g/cm3 (20 °C)
flash point   88 °C DIN 51758
autoignition temp.   225 °C DIN 51794
mp   -33 °C
LD 50 oral   LD50 Rat 1738 mg/kg
LD 50 dermal   LD50 Rat > 2000 mg/kg
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.942 - 0.946
  Identity (IR): passes test

Price and Availability

Safety & Documentation

Safety Information

HS Code 
2909 49 80
Hazard Statement(s) 
H302: Harmful if swallowed.
H319: Causes serious eye irritation.
H412: Harmful to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Categories of danger 
harmful, irritant, dangerous for the environment
R Phrase 
R 22-36-52/53
Harmful if swallowed.Irritating to eyes.Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 61
Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?