EMAIL THIS PAGE TO A FRIEND
8.40165 EMD Millipore

2-Oxazolidinone

2-Oxazolidinone for synthesis. CAS 497-25-6, molar mass 87.07 g/mol., for synthesis

  • CAS Number 497-25-6

  • Linear Formula C₃H₅NO₂

  • Molecular Weight 87.07 g/mol

  •  EC Number 207-840-9

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks
packaging   25 g in Glass bottle (8401650025)
mp   83 - 87 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 86 °C
  Melting range (upper value): ≤ 89 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 99 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?