• USA Home
  • 8.40171 - 3-Amino-4-methylbenzoic acid

EMAIL THIS PAGE TO A FRIEND
8.40171 EMD Millipore

3-Amino-4-methylbenzoic acid

3-Amino-4-methylbenzoic acid for synthesis. CAS 2458-12-0, molar mass 151.16 g/mol., for synthesis

Synonym: 3-Amino-p-toluic acid

  • CAS Number 2458-12-0

  • Linear Formula C₈H₉NO₂

  • Molecular Weight 151.16 g/mol

  •  EC Number 219-543-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 g in Glass bottle (8401710050)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 165 °C
  Melting range (upper value): ≤ 168 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 49 85
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?