EMAIL THIS PAGE TO A FRIEND
8.41001 EMD Millipore

5-Bromoindole

5-Bromoindole for synthesis. CAS No. 10075-50-0, EC Number 233-208-7., for synthesis

  • CAS Number 10075-50-0

  • Linear Formula C₈H₆BrN

  • Molecular Weight 196.04 g/mol

  •  EC Number 233-208-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8410010005)
solubility   126 mg/l (calculated)
mp   90 - 92 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 89 °C
  Melting range (upper value): ≤ 92 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?