• USA Home
  • 8.41002 - Potassium thioacetate

EMAIL THIS PAGE TO A FRIEND
8.41002 EMD Millipore

Potassium thioacetate

Potassium thioacetate for synthesis. CAS 10387-40-3, molar mass 114.21 g/mol., for synthesis

Synonym: Thioacetic acid potassium salt

  • CAS Number 10387-40-3

  • Linear Formula C₂H₃KOS

  • Molecular Weight 114.21 g/mol

  •  EC Number 233-848-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8410020025)
mp   220 - 225 °C
bulk density   500 kg/m3
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Precautionary Statement(s) 
P260: Do not breathe dust.
P262: Do not get in eyes, on skin, or on clothing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
UN 3335 , 9, III
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
S Phrase 
S 22-24/25
Do not breathe dust.Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?