• USA Home
  • 8.41008 - Methyl phenylacetate

EMAIL THIS PAGE TO A FRIEND
8.41008 EMD Millipore

Methyl phenylacetate

Methyl phenylacetate for synthesis. CAS 101-41-7, molar mass 150.18 g/mol., for synthesis

Synonym: Phenylacetic acid methyl ester

  • CAS Number 101-41-7

  • Linear Formula C₉H₁₀O₂

  • Molecular Weight 150.18 g/mol

  •  EC Number 202-940-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8410080250)
bp   93 - 95 °C (13.3 hPa)
vapor pressure   0.17 hPa (20 °C)
density   1.067 g/cm3 (20 °C)
flash point   85 °C
LD 50 oral   LD50 Rat 2550 mg/kg
LD 50 dermal   LD50 Rabbit 2400 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.065 - 1.070
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 10
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?