• USA Home
  • 8.41012 - 1-Methylpyrrol-2-carbaldehyde

EMAIL THIS PAGE TO A FRIEND
8.41012 EMD Millipore

1-Methylpyrrol-2-carbaldehyde

1-Methylpyrrol-2-carbaldehyde for synthesis. CAS 1192-58-1, molar mass 109.13 g/mol., for synthesis

Synonym: N-Methylpyrrole-2-carbaldehyde

  • CAS Number 1192-58-1

  • Linear Formula C₆H₇NO

  • Molecular Weight 109.13 g/mol

  •  EC Number 214-755-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8410120010)
bp   88 - 90 °C (29.3 hPa)
density   1.074 g/cm3 (20 °C)
flash point   72 °C
LD 50 oral   LD50 Rat 2204 mg/kg
LD 50 dermal   LD50 Rabbit > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.071 - 1.076
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?