EMAIL THIS PAGE TO A FRIEND
8.41015 EMD Millipore

Undecanal

Undecanal (stabilised) for synthesis. CAS 112-44-7, molar mass 170.29 g/mol., (stabilised) for synthesis

Synonym: Undecyl aldehyde, Hendecanal

  • CAS Number 112-44-7

  • Linear Formula C₁₁H₂₂O

  • Molecular Weight 170.29 g/mol

  •  EC Number 203-972-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8410150025)
bp   110 - 113 °C (16 hPa)
expl. lim.     0.7 %(V)
vapor pressure   0.05 hPa (20 °C)
density   0.827 g/cm3 (20 °C)
flash point   96 °C
mp   -2 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.825 - 0.835
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2912 19 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?