• USA Home
  • 8.41021 - Ethoxycarbonylmethylene-triphenylphosphorane

EMAIL THIS PAGE TO A FRIEND
8.41021 EMD Millipore

Ethoxycarbonylmethylene-triphenylphosphorane

Ethoxycarbonylmethylene-triphenylphosphorane for synthesis. CAS 1099-45-2, molar mass 348.38 g/mol., for synthesis

Synonym: (Triphenylphosphoranylidene)acetic acid ethyl ester, Ethyl triphenylphosphoranylidene-acetate

  • CAS Number 1099-45-2

  • Linear Formula C₂₂H₂₁O₂P

  • Molecular Weight 348.38 g/mol

  •  EC Number 214-151-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8410210010)
mp   125 - 127 °C
shipped in   ambient
storage temp.   Store at -20°C to 0°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 125 °C
  Melting range (upper value): ≤ 128 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 39 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?