• USA Home
  • 8.41024 - 4-Chloro-3-nitrobenzotrifluoride

EMAIL THIS PAGE TO A FRIEND
8.41024 EMD Millipore

4-Chloro-3-nitrobenzotrifluoride

4-Chloro-3-nitrobenzotrifluoride for synthesis. CAS 121-17-5, molar mass 225.55 g/mol., for synthesis

Synonym: 2-Chloro-5-(trifluoromethyl)nitrobenzene, CMNT

  • CAS Number 121-17-5

  • Linear Formula C₇H₃ClF₃NO₂

  • Molecular Weight 225.55 g/mol

  •  EC Number 204-451-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8410240100)
bp   108 - 109 °C (13 hPa)
density   1.535 g/cm3 (20 °C)
flash point   101 °C
mp   -3 °C
LD 50 oral   LD50 Rat 1075 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.533 - 1.538
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2904 99 00
Hazard Statement(s) 
H302: Harmful if swallowed.
H315: Causes skin irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2307 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2307 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2307 , 6.1, II, Marine Pollutant: P
Categories of danger 
harmful, irritant
R Phrase 
R 22-38
Harmful if swallowed.Irritating to skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?