• USA Home
  • 8.41026 - 4-Acetamidobenzenesulfonyl chloride

EMAIL THIS PAGE TO A FRIEND
8.41026 EMD Millipore

4-Acetamidobenzenesulfonyl chloride

4-Acetamidobenzenesulfonyl chloride for synthesis. CAS 121-60-8, chemical formula CH₃CONH(C₆H₄)SO₂CL., for synthesis

Synonym: N-Acetylsulfanilyl chloride

  • CAS Number 121-60-8

  • Linear Formula C₈H₈ClNO₃S

  • Molecular Weight 233.66 g/mol

  •  EC Number 204-485-1

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds More...
packaging   250 g in Glass bottle (8410260250)
flash point   >112 °C
mp   146 °C (decomposition)
LD 50 oral   LD50 Rat > 3200 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 98.0 %
  Melting range (lower value): ≥ 140 °C
  Melting range (upper value): ≤ 143 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 29 70
Hazard Statement(s) 
H314: Causes severe skin burns and eye damage.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 1759 , 8, II
Declaration (transport by air) IATA-DGR 
UN 1759 , 8, II
Declaration (transport by sea) IMDG-Code 
UN 1759 , 8, II
Categories of danger 
corrosive
R Phrase 
R 34-37
Causes burns.Irritating to respiratory system.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?