• USA Home
  • 8.41031 - Anthraquinone-2-sulfonic acid sodium salt

EMAIL THIS PAGE TO A FRIEND
8.41031 EMD Millipore

Anthraquinone-2-sulfonic acid sodium salt

Anthraquinone-2-sulfonic acid sodium salt for synthesis. CAS 131-08-8, molar mass 310.25 g/mol., for synthesis

Synonym: 9,19-Dihydro-9,10-dioxo-anthracene-2-sulfonic acid sodium salt

  • CAS Number 131-08-8

  • Linear Formula C₁₄H₇NaO₅S

  • Molecular Weight 310.25 g/mol

  •  EC Number 205-009-5

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds More...
packaging   100 g in Plastic bottle (8410310100)
mp   >320 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 97.0 % (a/a)
  Water (K. F.): ≤ 5.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 79 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?