• USA Home
  • 8.41037 - Decabromodiphenyl ether

EMAIL THIS PAGE TO A FRIEND
8.41037 EMD Millipore

Decabromodiphenyl ether

Decabromodiphenyl ether for synthesis. CAS 1163-19-5, molar mass 959.17 g/mol., for synthesis

Synonym: Perbromodiphenylether, Bis(pentabromophenyl)ether

  • CAS Number 1163-19-5

  • Linear Formula C₁₂Br₁₀O

  • Molecular Weight 959.17 g/mol

  •  EC Number 214-604-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8410370100)
vapor pressure   <1 Pa (21 °C)
solubility   <0.0001 mg/l
density   3.4 g/cm3
mp   304 - 307 °C
LD 50 oral   LD50 Rat > 2000 mg/kg
LD 50 dermal   LD50 Rabbit > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (ex Br): ≥ 97.0 %
  Melting range (lower value): ≥ 302 °C
  Melting range (upper value): ≤ 308 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 30 38
Hazard Statement(s) 
H413: May cause long lasting harmful effects to aquatic life.
Precautionary Statement(s) 
P273: Avoid release to the environment.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Categories of danger 
dangerous for the environment
R Phrase 
R 53
May cause long-term adverse effects in the aquatic environment.
S Phrase 
S 61
Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?