• USA Home
  • 8.41038 - Di-n-octyl phenylphosphonate

EMAIL THIS PAGE TO A FRIEND
8.41038 EMD Millipore

Di-n-octyl phenylphosphonate

Di-n-octyl phenylphosphonate for synthesis. CAS 1754-47-8, molar mass 382.52 g/mol., for synthesis

Synonym: DOPP

  • CAS Number 1754-47-8

  • Linear Formula C₂₂H₃₉O₃P

  • Molecular Weight 382.52 g/mol

  •  EC Number 217-142-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8410380005)
bp   219 - 220 °C (0.93 hPa)
density   0.932 g/cm3 (20 °C)
flash point   110 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.5 % (a/a)
  Density (d 20 °C/ 4 °C): 0.966 - 0.972
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 39 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?