• USA Home
  • 8.41039 - 3-Methoxyphenylacetic acid

EMAIL THIS PAGE TO A FRIEND
8.41039 EMD Millipore

3-Methoxyphenylacetic acid

3-Methoxyphenylacetic acid for synthesis. CAS 1798-09-0, molar mass 166.18 g/mol., for synthesis

  • CAS Number 1798-09-0

  • Linear Formula C₉H₁₀O₃

  • Molecular Weight 166.18 g/mol

  •  EC Number 217-282-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8410390010)
mp   68 - 72 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 97.5 %
  Melting range (lower value): ≥ 68 °C
  Melting range (upper value): ≤ 72 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 99 90
Hazard Statement(s) 
H318: Causes serious eye damage.
Precautionary Statement(s) 
P260: Do not breathe dust.
P280: Wear eye protection.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P313: Get medical advice/ attention.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 41
Risk of serious damage to eyes.
S Phrase 
S 22-26-39
Do not breathe dust.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear eye/face protection.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?