• USA Home
  • 8.41044 - 5-Aminopyrazole-4-carbonitrile

EMAIL THIS PAGE TO A FRIEND
8.41044 EMD Millipore

5-Aminopyrazole-4-carbonitrile

5-Aminopyrazole-4-carbonitrile for synthesis. CAS 16617-46-2, molar mass 108.10 g/mol., for synthesis

Synonym: 5-Amino-4-cyanopyrazole

  • CAS Number 16617-46-2

  • Linear Formula C₄H₄N₄

  • Molecular Weight 108.10 g/mol

  •  EC Number 240-665-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8410440001)
mp   173 - 175 °C
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (HPLC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 173 °C
  Melting range (upper value): ≤ 176 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 19 90
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 3439 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 3439 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 3439 , 6.1, III
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?