• USA Home
  • 8.41050 - Anthracene-9-methanol

EMAIL THIS PAGE TO A FRIEND
8.41050 EMD Millipore

Anthracene-9-methanol

Anthracene-9-methanol for synthesis. CAS 1468-95-7, EC Number 215-998-5, molar mass 208.25 g/mol., for synthesis

Synonym: 9-Anthrylmethanol, 9-(Hydroxymethyl)-anthracene

  • CAS Number 1468-95-7

  • Linear Formula C₁₅H₁₂O

  • Molecular Weight 208.25 g/mol

  •  EC Number 215-998-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Plastic bottle (8410500010)
mp   162 - 164 °C
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 159 °C
  Melting range (upper value): ≤ 163 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, III
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?