• USA Home
  • 8.41051 - Methyl 3-thienyl ketone

EMAIL THIS PAGE TO A FRIEND
8.41051 EMD Millipore

Methyl 3-thienyl ketone

Methyl 3-thienyl ketone for synthesis. CAS 1468-83-3, molar mass 126.17 g/mol., for synthesis

Synonym: 3-Acetylthiophene

  • CAS Number 1468-83-3

  • Linear Formula C₆H₆OS

  • Molecular Weight 126.17 g/mol

  •  EC Number 215-996-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8410510010)
mp   58 - 61 °C
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 58 °C
  Melting range (upper value): ≤ 61 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 99 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?