• USA Home
  • 8.41055 - 4,6-Dimethylpyrimidine

EMAIL THIS PAGE TO A FRIEND
8.41055 EMD Millipore

4,6-Dimethylpyrimidine

4,6-Dimethylpyrimidine for synthesis. CAS 1558-17-4, molar mass 108.14 g/mol., for synthesis

  • CAS Number 1558-17-4

  • Linear Formula C₆H₈N₂

  • Molecular Weight 108.14 g/mol

  •  EC Number 216-314-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8410550010)
bp   153 - 155 °C (1013 hPa)
density   0.98 g/cm3 (20 °C)
flash point   46 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.988 - 0.995
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 59 95
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, II
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?