• USA Home
  • 8.41057 - 1-Bromo-4-ethylbenzene

EMAIL THIS PAGE TO A FRIEND
8.41057 EMD Millipore

1-Bromo-4-ethylbenzene

1-Bromo-4-ethylbenzene for synthesis. CAS 1585-07-5, molar mass 185.06 g/mol., for synthesis

  • CAS Number 1585-07-5

  • Linear Formula C₈H₉Br

  • Molecular Weight 185.06 g/mol

  •  EC Number 216-439-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8410570010)
bp   203 - 204 °C (1013 hPa)
vapor pressure   0.9 hPa (25 °C)
density   1.34 g/cm3 (20 °C)
flash point   63 °C
mp   -44 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.5 % (a/a)
  Density (d 20 °C/ 4 °C): 1.342 - 1.344
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Hazard Statement(s) 
H315: Causes skin irritation.
H318: Causes serious eye damage.
Precautionary Statement(s) 
P280: Wear eye protection.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P313: Get medical advice/ attention.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 38-41
Irritating to skin.Risk of serious damage to eyes.
S Phrase 
S 26-39
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear eye/face protection.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?