• USA Home
  • 8.41058 - Indole-5-carbonitrile

EMAIL THIS PAGE TO A FRIEND
8.41058 EMD Millipore

Indole-5-carbonitrile

Indole-5-carbonitrile for synthesis. CAS 15861-24-2, molar mass 142.16 g/mol., for synthesis

  • CAS Number 15861-24-2

  • Linear Formula C₉H₆N₂

  • Molecular Weight 142.16 g/mol

  •  EC Number 239-986-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8410580001)
mp   106 - 108 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 105 °C
  Melting range (upper value): ≤ 108 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
RTECS 
NL5993120
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?