EMAIL THIS PAGE TO A FRIEND
8.41059 EMD Millipore

(-)-Menthone

(-)-Menthone for synthesis. CAS 14073-97-3, molar mass 154.25 g/mol., for synthesis

Synonym: (2S,5R)-(-)-2-Isopropyl 5-methylcyclohexanone, 2S-trans-5-Methyl-2-(1-methylethyl)cyclohexanone

  • CAS Number 14073-97-3

  • Linear Formula C₁₀H₁₈O

  • Molecular Weight 154.25 g/mol

  •  EC Number 237-926-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8410590025)
density   0.894 g/cm3 (20 °C)
flash point   72 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.5 % (a/a)
  Density (d 20 °C/ 4 °C): 0.890 - 0.895
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?