• USA Home
  • 8.41061 - 4-Acetylbenzonitrile

EMAIL THIS PAGE TO A FRIEND
8.41061 EMD Millipore

4-Acetylbenzonitrile

4-Acetylbenzonitrile for synthesis. CAS 1443-80-7, molar mass 145.16 g/mol., for synthesis

Synonym: 4-Cyanoacetophenone

  • CAS Number 1443-80-7

  • Linear Formula C₉H₇NO

  • Molecular Weight 145.16 g/mol

  •  EC Number 215-885-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8410610025)
mp   57 - 58 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 56 °C
  Melting range (upper value): ≤ 58 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Hazard Statement(s) 
H302 + H312: Harmful if swallowed or in contact with skin
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Categories of danger 
harmful
R Phrase 
R 21/22
Harmful in contact with skin and if swallowed.
S Phrase 
S 23
Do not breathe vapour.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?