• USA Home
  • 8.41064 - Diphenylphosphinic acid

EMAIL THIS PAGE TO A FRIEND
8.41064 EMD Millipore

Diphenylphosphinic acid

Diphenylphosphinic acid for synthesis. CAS 1707-03-5, molar mass 218.19 g/mol., for synthesis

  • CAS Number 1707-03-5

  • Linear Formula C₁₂H₁₁O₂P

  • Molecular Weight 218.19 g/mol

  •  EC Number 216-948-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8410640005)
mp   193 - 195 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Melting range (lower value): ≥ 192 °C
  Melting range (upper value): ≤ 196 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 39 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?