• USA Home
  • 8.41067 - 5-Amino-2-fluorobenzotrifluoride

EMAIL THIS PAGE TO A FRIEND
8.41067 EMD Millipore

5-Amino-2-fluorobenzotrifluoride

5-Amino-2-fluorobenzotrifluoride for synthesis. CAS 2357-47-3, chemical formula FC₆H₃(CF₃)NH₂., for synthesis

Synonym: 4-Fluoro-3-(trifluoromethyl)aniline

  • CAS Number 2357-47-3

  • Linear Formula C₇H₅F₄N

  • Molecular Weight 179.11 g/mol

  •  EC Number 219-095-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 ml in Glass bottle (8410670001)
bp   207 - 208 °C (1013 hPa)
density   1.393 g/cm3 (20 °C)
flash point   75 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.411 - 1.415
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 43 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?