• USA Home
  • 8.41071 - 2,4-Thiazolidinedione

EMAIL THIS PAGE TO A FRIEND
8.41071 EMD Millipore

2,4-Thiazolidinedione

2,4-Thiazolidinedione for synthesis. CAS 2295-31-0, molar mass 117.13 g/mol., for synthesis

  • CAS Number 2295-31-0

  • Linear Formula C₃H₃NO₂S

  • Molecular Weight 117.13 g/mol

  •  EC Number 218-941-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8410710025)
mp   123 - 126 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 124 °C
  Melting range (upper value): ≤ 127 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 10 00
RTECS 
XJ5775000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?