• USA Home
  • 8.41074 - 3-Amino-1-phenyl-butane

EMAIL THIS PAGE TO A FRIEND
8.41074 EMD Millipore

3-Amino-1-phenyl-butane

3-Amino-1-phenyl-butane for synthesis. CAS 22374-89-6, pH 11.6 (10 g/l, H₂O, 20 °C)., for synthesis

Synonym: 4-Phenyl-2-butylamine, 1-Methyl-3-phenylpropylamine

  • CAS Number 22374-89-6

  • Linear Formula C₁₀H₁₅N

  • Molecular Weight 149.23 g/mol

  •  EC Number 244-942-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8410740010)
density   0.928 g/cm3 (20 °C)
flash point   97 °C
autoignition temp.   330 °C
mp   <=-70 °C
pH   11.6 (10 g/l, H₂O, 20 °C)
LD 50 oral   LD50 Rat 165 mg/kg
LD 50 dermal   LD50 Rabbit 360 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.924 - 0.928
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 49 00
Hazard Statement(s) 
H301 + H311: Toxic if swallowed or in contact with skin
H314: Causes severe skin burns and eye damage.
H412: Harmful to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1A Combustible substances, toxic
WGK 
WGK 2 water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2922 , 8 (6.1), II
Declaration (transport by air) IATA-DGR 
UN 2922 , 8 (6.1), II
Declaration (transport by sea) IMDG-Code 
UN 2922 , 8 (6.1), II, Segregation Group: 18 (Alkalis)
Categories of danger 
toxic, corrosive, dangerous for the environment
R Phrase 
R 24/25-34-52/53
Toxic in contact with skin and if swallowed.Causes burns.Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 26-36/37/39-45-61
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?