• USA Home
  • 8.41076 - 1,2,3,4-Tetrahydro-6,7-dimethoxy-isoquinoline hydrochloride

EMAIL THIS PAGE TO A FRIEND
8.41076 EMD Millipore

1,2,3,4-Tetrahydro-6,7-dimethoxy-isoquinoline hydrochloride

for synthesis

  • CAS Number 2328-12-3

  • Linear Formula C₁₁H₁₆ClNO₂

  • Molecular Weight 229.7 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Plastic bottle (8410760005)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 97.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 49 90
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?