EMAIL THIS PAGE TO A FRIEND
8.41078 EMD Millipore

5-Indanole

5-Indanole for synthesis. CAS 1470-94-6, molar mass 134.18 g/mol., for synthesis

Synonym: 5-Hydroxyindan, 5-Hydroxyhydrinden

  • CAS Number 1470-94-6

  • Linear Formula C₉H₁₀O

  • Molecular Weight 134.18 g/mol

  •  EC Number 216-006-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8410780025)
solubility   6.3 g/l
flash point   130 °C
autoignition temp.   500 °C
mp   50 - 53 °C
LD 50 oral   LD50 Rat 3250 mg/kg
LD 50 dermal   LD50 Rabbit 450 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 50 °C
  Melting range (upper value): ≤ 53 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2907 19 90
Hazard Statement(s) 
H311: Toxic in contact with skin.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, III
R Phrase 
R 21
Harmful in contact with skin.
S Phrase 
S 36/37
Wear suitable protective clothing and gloves.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?