• USA Home
  • 8.41080 - 3,5-Diamino-1,2,4-triazole

EMAIL THIS PAGE TO A FRIEND
8.41080 EMD Millipore

3,5-Diamino-1,2,4-triazole

3,5-Diamino-1,2,4-triazole for synthesis. CAS 1455-77-2, molar mass 99.09 g/mol., for synthesis

Synonym: Guanazole

  • CAS Number 1455-77-2

  • Linear Formula C₂H₅N₅

  • Molecular Weight 99.09 g/mol

  •  EC Number 215-937-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Plastic bottle (8410800010)
mp   205 °C (decomposition)
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 97.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?